siRNA Details
| Gene Symbol: 1700093K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110133.1 | NP_001103603.1 | 4 | 833 | ||
| NM_026105.3 | NP_080381.2 | 4 | 515 | |||
| GenBank mRNA | AK006386.1 | - | 1 | 19 | ||
| AK007052.1 | - | 4 | 421 | |||
| AK007139.1 | - | 3 | 272 | |||
| BC049567.1 | - | 4 | 515 | 6773879 | ||

1700093K21Rik
Genomic Map