siRNA Details
| Gene Symbol: 1700093K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110133.1 | NP_001103603.1 | 3, 4 | 737 | ||
| NM_026105.3 | NP_080381.2 | 3, 4 | 419 | |||
| GenBank mRNA | AK007052.1 | - | 3, 4 | 325 | ||
| AK007139.1 | - | 2, 3 | 176 | |||
| BC049567.1 | - | 3, 4 | 419 | 6773879 | ||

1700093K21Rik
Genomic Map