siRNA Details
| Gene Symbol: Brd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113573.1 | NP_001107045.1 | 7 | 1322 | ||
| NM_001113574.1 | NP_001107046.1 | 7 | 1322 | |||
| NM_023336.4 | NP_075825.3 | 7 | 1482 | |||
| GenBank mRNA | AB206708.2 | - | 6 | 1133 | ||
| AF269193.1 | - | 7 | 1270 | |||
| AK037435.1 | - | 7 | 1304 | |||
| AK076472.1 | - | 7 | 1253 | |||
| AK076924.1 | - | 7 | 1401 | |||
| AK081471.1 | - | 7 | 1303 | |||
| AK136007.1 | - | 7 | 1282 | |||
| AK136429.1 | - | 7 | 1253 | |||
| AK159827.1 | - | 7 | 1308 | |||
| AK160752.1 | - | 7 | 1281 | |||
| AK161164.1 | - | 7 | 1278 | |||
| AK167573.1 | - | 7 | 1280 | |||
| AY513269.1 | - | 6 | 1148 | |||
| BC004832.1 | - | 7 | 1267 | |||
| BC031536.1 | - | 7 | 1296 | 4015879 | ||

Brd3
Genomic Map