siRNA Details
| Gene Symbol: Bag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026121.3 | NP_080397.1 | 3 | 442 | ||
| GenBank mRNA | AF332863.1 | - | 3 | 435 | ||
| AK010765.1 | - | 3 | 433 | |||
| AK136899.1 | - | 3 | 432 | |||
| AK160316.1 | - | 3 | 434 | |||
| AK162658.1 | - | 3 | 429 | |||
| BC037239.1 | - | 3 | 440 | |||
| BC058518.1 | - | 3 | 425 | 5717401 | ||
| BC138201.1 | - | 3 | 520 | 8861220 | ||
| BC145986.1 | - | 3 | 520 | 40131289 | ||
| BU939693.1 | - | 447 | ||||

Bag4
Genomic Map