siRNA Details
Gene Symbol: Gm17484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090261.1 | - | 2 | 644 |
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026121.3 | NP_080397.1 | 5 | 1277 | ||
GenBank mRNA | AF332863.1 | - | 5 | 1270 | ||
AK010765.1 | - | 5 | 1268 | |||
AK136899.1 | - | 5 | 1267 | |||
AK160316.1 | - | 5 | 1269 | |||
AK162658.1 | - | 5 | 1264 | |||
BC009102.1 | - | 1 | 311 | |||
BC037239.1 | - | 5 | 1275 | |||
BC058518.1 | - | 5 | 1260 | 5717401 | ||
BC138201.1 | - | 5 | 1355 | 8861220 | ||
BC145986.1 | - | 5 | 1355 | 40131289 |
