siRNA Details
| Gene Symbol: Rnmtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183263.5 | NP_899086.2 | 4 | 889 | ||
| GenBank mRNA | AK012200.1 | - | 4 | 864 | ||
| AK014739.1 | - | 4 | 855 | |||
| AK147805.1 | - | 4 | 870 | |||
| BC038288.1 | - | 4 | 838 | |||
| BC090975.1 | - | 4 | 861 | 6595681 | ||
| BC115622.1 | - | 4 | 615 | 40102799 | ||
| BC115623.1 | - | 4 | 615 | 40102801 | ||

Rnmtl1
Genomic Map