siRNA Details
| Gene Symbol: Fundc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026126.4 | NP_080402.3 | 1, 2 | 55 | ||
| GenBank mRNA | AK013460.1 | - | 1, 2 | 33 | ||
| AK014705.1 | - | 1, 2 | 32 | |||
| AK031214.1 | - | 1, 2 | 32 | |||
| AY234858.1 | - | 1, 2 | 21 | |||
| BC042794.1 | - | 12 | ||||
| BC049566.1 | - | 1, 2 | 30 | 6742356 | ||
| BC061464.1 | - | 1, 2 | 25 | 6819539 | ||
| BY124891.1 | - | 1, 2 | 55 | |||

Fundc2
Genomic Map