siRNA Details
| Gene Symbol: Nvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026171.1 | NP_080447.1 | 19 | 2308 | ||
| GenBank mRNA | AK004676.1 | - | 19 | 2340 | ||
| AK054502.1 | - | 14 | 1787 | |||
| AK134620.1 | - | 2 | 154 | |||
| AK140509.1 | - | 19 | 2339 | |||
| BC031847.1 | - | 19 | 2291 | 3964470 | ||

Nvl
Genomic Map