siRNA Details
| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026172.3 | NP_080448.1 | 3 | 458 | ||
| GenBank mRNA | AK002756.1 | - | 3 | 338 | ||
| AK004725.2 | - | 3 | 325 | |||
| AK015692.1 | - | 3 | 329 | |||
| AK146003.1 | - | 3 | 329 | |||
| AK168490.1 | - | 3 | 326 | |||
| BC046972.1 | - | 3 | 458 | 6469277 | ||
| CT010405.1 | - | 3 | 302 | |||

Decr1
Genomic Map