siRNA Details
| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026172.3 | NP_080448.1 | 9 | 1084 | ||
| GenBank mRNA | AK002756.1 | - | 9 | 964 | ||
| AK004725.2 | - | 9 | 951 | |||
| AK010473.1 | - | 9 | 945 | |||
| AK015692.1 | - | 9 | 955 | |||
| AK146003.1 | - | 9 | 955 | |||
| AK168490.1 | - | 9 | 952 | |||
| BC046972.1 | - | 9 | 1084 | 6469277 | ||
| CT010405.1 | - | 9 | 928 | |||

Decr1
Genomic Map