siRNA Details
| Gene Symbol: Gpatch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026181.1 | NP_080457.1 | 9 | 1048 | ||
| GenBank mRNA | AK004868.1 | - | 9 | 1048 | ||
| AK014367.1 | - | 9 | 1027 | |||
| AK154571.1 | - | 9 | 1029 | |||
| BC026394.1 | - | 3 | 214 | |||
| BC100577.1 | - | 9 | 1023 | |||
| BC125584.1 | - | 9 | 1039 | 40130099 | ||
| BC132155.1 | - | 9 | 1039 | 40130432 | ||

Gpatch1
Genomic Map