siRNA Details
| Gene Symbol: Eepd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026189.3 | NP_080465.3 | 7 | 1882 | ||
| GenBank mRNA | AK009180.2 | - | 7 | 1877 | ||
| AK019086.1 | - | 7 | 1882 | |||
| AK168564.1 | - | 7 | 1883 | |||
| AK173239.1 | - | 7 | 1932 | |||
| BC025571.1 | - | 7 | 1946 | 5323841 | ||
| CK619108.1 | - | 6 | 609 | |||

Eepd1
Genomic Map