siRNA Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026194.4 | NP_080470.2 | 9 | 939 | ||
| GenBank mRNA | AK007993.2 | - | 9 | 884 | ||
| AK013382.1 | - | 2 | 202 | |||
| AK032663.1 | - | 9 | 875 | |||
| AK129211.1 | - | 8 | 852 | |||
| AK132697.1 | - | 9 | 903 | |||
| BC058809.1 | - | 8 | 2263 | |||
| BC080203.1 | - | 9 | 917 | |||
| BC099529.1 | - | 9 | 926 | |||
| BC132195.1 | - | 9 | 891 | 40130472 | ||
| BC138153.1 | - | 9 | 891 | 8861172 | ||

Ufl1
Genomic Map