siRNA Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026218.2 | NP_080494.1 | 5, 6 | 805 | ||
| GenBank mRNA | AK004662.1 | - | 5, 6 | 789 | ||
| AK005320.1 | - | 5, 6 | 782 | |||
| AK050102.1 | - | 5, 6 | 806 | |||
| AK159805.1 | - | 5, 6 | 754 | |||

Fgfr1op2
Genomic Map