siRNA Details
| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014974.1 | NP_001014974.1 | 12, 13 | 2879 | ||
| GenBank mRNA | AK129075.1 | - | 10, 11 | 1706 | ||
| AM690747.1 | - | 10, 11 | 2482 | |||
| BC085151.1 | - | 12, 13 | 2879 | 30534216 | ||

Ttll4
Genomic Map