siRNA Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026231.2 | NP_080507.2 | 3 | 855 | ||
| GenBank mRNA | AK020021.1 | - | 2 | 465 | ||
| AK029724.1 | - | 3 | 854 | |||
| AK033409.1 | - | 3 | 893 | |||
| AK037169.1 | - | 3 | 894 | |||
| AK044441.1 | - | 3 | 863 | |||
| BC047136.1 | - | 3 | 855 | 6311866 | ||
| BC061240.1 | - | 3 | 851 | 6743615 | ||

Gstcd
Genomic Map