siRNA Details
Gene Symbol: Mgat4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026233.2 | NP_080509.2 | 1, 2 | 97 | ||
GenBank mRNA | AK016272.1 | - | 1, 2 | 106 | ||
HM067443.1 | - | 1, 2 | 221 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgat4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026233.2 | NP_080509.2 | 1, 2 | 97 | ||
GenBank mRNA | AK016272.1 | - | 1, 2 | 106 | ||
HM067443.1 | - | 1, 2 | 221 |