siRNA Details
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026235.4 | NP_080511.2 | 3 | 894 | ||
GenBank mRNA | AK017372.1 | - | 3 | 899 | ||
AK020966.1 | - | 3 | 831 | |||
AK035809.1 | - | 471 | ||||
AK039614.1 | - | 3 | 877 | |||
AK042616.1 | - | 3 | 826 | |||
AK047452.1 | - | 1 | 198 | |||
AK087521.1 | - | 3 | 897 | |||
BC090615.1 | - | 3 | 1026 | 6809572 |
