siRNA Details
| Gene Symbol: Larp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026235.4 | NP_080511.2 | 3 | 738 | ||
| GenBank mRNA | AK017372.1 | - | 3 | 743 | ||
| AK020966.1 | - | 3 | 675 | |||
| AK035809.1 | - | 315 | ||||
| AK039614.1 | - | 3 | 721 | |||
| AK042616.1 | - | 3 | 670 | |||
| AK047452.1 | - | 1 | 42 | |||
| AK087521.1 | - | 3 | 741 | |||
| BC090615.1 | - | 3 | 870 | 6809572 | ||

Larp6
Genomic Map