siRNA Details
| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026254.2 | NP_080530.2 | 4 | 484 | ||
| GenBank mRNA | AK155142.1 | - | 4 | 505 | ||
| AK171591.1 | - | 4 | 392 | |||
| BC031706.1 | - | 4 | 484 | 4194266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d23| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026254.2 | NP_080530.2 | 4 | 484 | ||
| GenBank mRNA | AK155142.1 | - | 4 | 505 | ||
| AK171591.1 | - | 4 | 392 | |||
| BC031706.1 | - | 4 | 484 | 4194266 | ||