siRNA Details
| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026254.2 | NP_080530.2 | 18 | 2100 | ||
| GenBank mRNA | AK015437.1 | - | 9 | 932 | ||
| AK155142.1 | - | 18 | 2121 | |||
| BC019762.1 | - | 8 | 838 | |||
| BC031706.1 | - | 18 | 2100 | 4194266 | ||

Tbc1d23
Genomic Map