siRNA Details
| Gene Symbol: Tctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026260.2 | NP_080536.2 | 7 | 908 | ||
| GenBank mRNA | AK015861.1 | - | 7 | 896 | ||
| AK016982.1 | - | 7 | 894 | |||
| AK030156.1 | - | 7 | 894 | |||
| BC027335.1 | - | 7 | 908 | 3710046 | ||
| BC080805.1 | - | 7 | 883 | 5352578 | ||
| DQ278871.1 | - | 7 | 925 | |||

Tctn3
Genomic Map