siRNA Details
| Gene Symbol: Tespa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183264.4 | NP_899087.2 | 9 | 1279 | ||
| GenBank mRNA | AK017900.1 | - | 8 | 1018 | ||
| AK156313.1 | - | 9 | 1255 | |||
| BC064065.1 | - | 5 | 865 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tespa1| Gene Symbol: Tespa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183264.4 | NP_899087.2 | 9 | 1279 | ||
| GenBank mRNA | AK017900.1 | - | 8 | 1018 | ||
| AK156313.1 | - | 9 | 1255 | |||
| BC064065.1 | - | 5 | 865 | |||