siRNA Details
| Gene Symbol: Necap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026267.2 | NP_080543.2 | 5, 6 | 493 | ||
| GenBank mRNA | AK004805.1 | - | 5, 6 | 494 | ||
| AK005263.1 | - | 5, 6 | 484 | |||
| AK081598.1 | - | 5, 6 | 494 | |||
| AK168500.1 | - | 5, 6 | 494 | |||
| AK211496.1 | - | 2, 3 | 182 | |||
| AK220236.1 | - | 5, 6 | 479 | |||
| BC011466.1 | - | 5, 6 | 492 | 4013043 | ||
| BC119133.1 | - | 5, 6 | 505 | 8733882 | ||
| BC120841.1 | - | 5, 6 | 576 | 40129764 | ||

Necap1
Genomic Map