siRNA Details
| Gene Symbol: Narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026272.3 | NP_080548.3 | 7 | 827 | ||
| GenBank mRNA | AK013432.1 | - | 7 | 786 | ||
| AK076232.1 | - | 7 | 810 | |||
| AK081305.1 | - | 7 | 811 | |||
| BC016090.1 | - | 7 | 766 | 4503713 | ||

Narf
Genomic Map