siRNA Details
| Gene Symbol: Sdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023374.3 | NP_075863.2 | 2 | 324 | ||
| GenBank mRNA | AK003052.1 | - | 2 | 218 | ||
| AK003533.1 | - | 2 | 208 | |||
| AK009660.1 | - | 2 | 218 | |||
| AK153045.1 | - | 2 | 201 | |||
| AK169252.1 | - | 2 | 215 | |||
| AK169839.1 | - | 2 | 217 | |||
| BC013509.1 | - | 2 | 219 | 4225025 | ||
| BC018580.1 | - | 2 | 183 | |||
| BC051934.1 | - | 2 | 324 | 5688898 | ||
| DQ403000.1 | - | 1 | 115 | |||

Sdhb
Genomic Map