siRNA Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026312.5 | NP_080588.1 | 6 | 605 | ||
| GenBank mRNA | AK007872.1 | - | 5 | 474 | ||
| AK011610.1 | - | 6 | 569 | |||
| AK030507.1 | - | 6 | 587 | |||
| BC035042.1 | - | 6 | 569 | 1224959 | ||
| BC044719.1 | - | 6 | 553 | 6394829 | ||
| BC061070.1 | - | 6 | 566 | 6594922 | ||

Pbdc1
Genomic Map