siRNA Details
Gene Symbol: Pbdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281871.1 | NP_001268800.1 | 4 | 329 | ||
NM_026312.5 | NP_080588.1 | 4 | 329 | |||
GenBank mRNA | AK007655.1 | - | 4 | 311 | ||
AK007872.1 | - | 3 | 198 | |||
AK011610.1 | - | 4 | 293 | |||
AK030507.1 | - | 4 | 311 | |||
AK076889.1 | - | 4 | 311 | |||
AK157432.1 | - | 2 | 2169 | |||
BC035042.1 | - | 4 | 293 | 1224959 | ||
BC044719.1 | - | 4 | 277 | 6394829 | ||
BC061070.1 | - | 4 | 290 | 6594922 |
