siRNA Details
| Gene Symbol: Rnf149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033135.3 | NP_001028307.2 | 6 | 1076 | ||
| GenBank mRNA | AK155360.1 | - | 6 | 1135 | ||
| AY155439.1 | - | 6 | 668 | |||
| BC115968.1 | - | 7 | 656 | 40107454 | ||
| CR759684.1 | - | 5 | 454 | |||

Rnf149
Genomic Map