siRNA Details
| Gene Symbol: Stx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026343.2 | NP_080619.2 | 4 | 475 | ||
| GenBank mRNA | AK011400.1 | - | 4 | 469 | ||
| AK014718.1 | - | 3 | 414 | |||
| AK018526.1 | - | 4 | 465 | |||
| AK034718.1 | - | 4 | 464 | |||
| AK135240.1 | - | 4 | 471 | |||
| AK154503.1 | - | 4 | 476 | |||
| BC028639.1 | - | 4 | 462 | 1243532 | ||

Stx17
Genomic Map