siRNA Details
| Gene Symbol: Ccdc130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294281.1 | NP_001281210.1 | 7 | 663 | ||
| NM_026350.3 | NP_080626.1 | 7 | 663 | |||
| GenBank mRNA | AK015911.1 | - | 7 | 622 | ||
| BC008614.1 | - | 2 | 276 | |||
| BC021321.1 | - | 6 | 619 | 5041100 | ||

Ccdc130
Genomic Map