siRNA Details
| Gene Symbol: Mgarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026358.3 | NP_080634.2 | 1, 2 | 128 | ||
| GenBank mRNA | AF228502.1 | - | 1, 2 | 107 | ||
| AK006339.1 | - | 1, 2 | 90 | |||
| AK016342.1 | - | 1, 2 | 101 | |||
| AK151401.1 | - | 1, 2 | 122 | |||
| BC012511.1 | - | 1, 2 | 121 | 4500810 | ||
| BC049556.1 | - | 1, 2 | 90 | 6772062 | ||
| BY328038.1 | - | 1, 2 | 128 | |||

Mgarp
Genomic Map