siRNA Details
| Gene Symbol: Eqtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290623.1 | NP_001277552.1 | 2 | 240 | ||
| NM_027089.4 | NP_081365.2 | 2 | 240 | |||
| GenBank mRNA | AB438105.1 | - | 2 | 149 | ||
| AB438106.1 | - | 2 | 149 | |||
| AK006446.1 | - | 2 | 191 | |||
| AK016313.1 | - | 2 | 208 | |||
| AK016738.1 | - | 2 | 211 | |||
| BB572592.1 | - | 2 | 241 | |||
| BC089517.1 | - | 2 | 207 | 6740450 | ||
| BY716419.1 | - | 2 | 207 | |||
| DQ336137.1 | - | 2 | 191 | |||
| EU828781.1 | - | 2 | 225 | |||
| EU828782.1 | - | 2 | 225 | |||

Eqtn
Genomic Map