siRNA Details
| Gene Symbol: Rtp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023386.5 | NP_075875.3 | 2 | 299 | ||
| GenBank mRNA | AJ251364.1 | - | 2 | 294 | ||
| AK007477.1 | - | 2 | 277 | |||
| AK018021.1 | - | 2 | 292 | |||
| AY562228.1 | - | 2 | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtp4| Gene Symbol: Rtp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023386.5 | NP_075875.3 | 2 | 299 | ||
| GenBank mRNA | AJ251364.1 | - | 2 | 294 | ||
| AK007477.1 | - | 2 | 277 | |||
| AK018021.1 | - | 2 | 292 | |||
| AY562228.1 | - | 2 | 229 | |||