siRNA Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161818.1 | NP_001155290.1 | 7 | 1313 | ||
| NM_144519.4 | NP_653102.1 | 7 | 1414 | |||
| GenBank mRNA | AK049934.1 | - | 7 | 1293 | ||
| AK076145.1 | - | 7 | 1415 | |||
| AK076837.1 | - | 7 | 1290 | |||
| AK078000.1 | - | 7 | 1314 | |||
| AY736186.1 | - | 4 | 999 | |||
| BC003941.1 | - | 5 | 1063 | 3487151 | ||
| BC042660.1 | - | 1 | 655 | |||

Zfp639
Genomic Map