siRNA Details
| Gene Symbol: Plxnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026376.3 | NP_080652.2 | 35 | 5826 | ||
| GenBank mRNA | AK129175.1 | - | 35 | 5091 | ||
| AK147513.1 | - | 35 | 5827 | |||
| AY688678.1 | - | 35 | 5786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxnd1| Gene Symbol: Plxnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026376.3 | NP_080652.2 | 35 | 5826 | ||
| GenBank mRNA | AK129175.1 | - | 35 | 5091 | ||
| AK147513.1 | - | 35 | 5827 | |||
| AY688678.1 | - | 35 | 5786 | |||