siRNA Details
Gene Symbol: Brix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026396.3 | NP_080672.3 | 9 | 790 | ||
GenBank mRNA | AK002985.1 | - | 9 | 677 | ||
AK088313.1 | - | 9 | 778 | |||
AK168325.1 | - | 9 | 763 | |||
BC014832.1 | - | 9 | 760 | 4484024 | ||
CF749026.1 | - | 2 | 192 |
