siRNA Details
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029573.2 | NP_083849.1 | 9 | 854 | ||
GenBank mRNA | AK003393.1 | - | 6 | 747 | ||
AK010065.1 | - | 9 | 878 | |||
AK032787.1 | - | 9 | 878 | |||
AK047951.1 | - | 9 | 876 | |||
AK117103.1 | - | 9 | 890 | |||
AK146717.1 | - | 6 | 718 | |||
AK150618.1 | - | 9 | 885 | |||
AK151304.1 | - | 9 | 879 | |||
AK152353.1 | - | 9 | 888 | |||
AK153459.1 | - | 9 | 879 | |||
AK158646.1 | - | 9 | 1032 | |||
AK159051.1 | - | 9 | 951 | |||
AK164038.1 | - | 10 | 981 | |||
AK167822.1 | - | 9 | 884 | |||
AK168034.1 | - | 9 | 876 | |||
AK168049.1 | - | 9 | 879 | |||
AK168149.1 | - | 9 | 885 | |||
AK169152.1 | - | 9 | 852 | |||
BC034273.1 | - | 9 | 844 | 4166193 | ||
BC049956.1 | - | 9 | 854 | 4212315 |
