siRNA Details
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029573.2 | NP_083849.1 | 3 | 128 | ||
GenBank mRNA | AK010065.1 | - | 3 | 152 | ||
AK032787.1 | - | 3 | 152 | |||
AK047951.1 | - | 3 | 150 | |||
AK117103.1 | - | 3 | 162 | |||
AK150618.1 | - | 3 | 159 | |||
AK151304.1 | - | 3 | 153 | |||
AK152353.1 | - | 3 | 162 | |||
AK153459.1 | - | 3 | 153 | |||
AK158646.1 | - | 3 | 306 | |||
AK159051.1 | - | 3 | 225 | |||
AK159666.1 | - | 3 | 159 | |||
AK160054.1 | - | 3 | 159 | |||
AK164038.1 | - | 4 | 254 | |||
AK167822.1 | - | 3 | 159 | |||
AK168034.1 | - | 3 | 150 | |||
AK168049.1 | - | 3 | 153 | |||
AK168149.1 | - | 3 | 159 | |||
AK169152.1 | - | 3 | 126 | |||
BC034273.1 | - | 3 | 118 | 4166193 | ||
BC049956.1 | - | 3 | 128 | 4212315 |
