siRNA Details
| Gene Symbol: Slco6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006249.1 | - | 1 | 101 | ||
| AK016463.1 | - | 3 | 847 | |||
| AY461596.1 | - | 3 | 853 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco6b1| Gene Symbol: Slco6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006249.1 | - | 1 | 101 | ||
| AK016463.1 | - | 3 | 847 | |||
| AY461596.1 | - | 3 | 853 | |||