siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026430.3 | NP_080706.1 | 13 | 998 | ||
| GenBank mRNA | AF399958.1 | - | 13 | 998 | ||
| AK005536.1 | - | 13 | 938 | |||
| AK075806.1 | - | 13 | 1050 | |||
| AK076166.1 | - | 12 | 1039 | |||
| AK152376.1 | - | 13 | 1053 | |||
| AK177105.1 | - | 2 | 96 | |||
| AK184776.1 | - | 2 | 96 | |||
| AK186705.1 | - | 2 | 96 | |||
| BC037049.1 | - | 13 | 1044 | 4935237 | ||

Uxs1
Genomic Map