siRNA Details
| Gene Symbol: Ppa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026438.4 | NP_080714.2 | 6 | 572 | ||
| GenBank mRNA | AK008575.1 | - | 6 | 573 | ||
| AK020291.1 | - | 6 | 577 | |||
| AK088397.1 | - | 6 | 568 | |||
| AK145640.1 | - | 6 | 622 | |||
| AK146300.1 | - | 6 | 535 | |||
| AK151512.1 | - | 6 | 563 | |||
| AK219020.1 | - | 3 | 231 | |||
| BC010468.1 | - | 6 | 506 | 3585780 | ||
| CT010203.1 | - | 6 | 465 | |||

Ppa1
Genomic Map