siRNA Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026447.4 | NP_080723.3 | 8 | 1077 | ||
| NM_198931.3 | NP_945149.2 | 8 | 1081 | |||
| GenBank mRNA | AB474373.1 | - | 8 | 1116 | ||
| AK013149.1 | - | 8 | 1010 | |||
| AK017245.1 | - | 5 | 414 | |||
| AK046387.1 | - | 976 | ||||
| AK087999.1 | - | 8 | 1115 | |||
| AK216174.1 | - | 3 | 179 | |||
| AY332616.1 | - | 8 | 1084 | |||
| BC058248.1 | - | 8 | 1003 | |||

Ppm1m
Genomic Map