siRNA Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026470.3 | NP_080746.3 | 8, 9 | 1064 | ||
| GenBank mRNA | AF032967.1 | - | 2, 3 | 371 | ||
| AK005819.1 | - | 3, 4 | 451 | |||
| AK006861.1 | - | 3, 4 | 436 | |||
| AK051574.1 | - | 8, 9 | 1065 | |||
| AK132771.1 | - | 8, 9 | 1038 | |||
| AK183508.1 | - | 2, 3 | 145 | |||
| BC028429.1 | - | 3, 4 | 464 | |||
| BC040024.1 | - | 3, 4 | 464 | |||

Spata6
Genomic Map