siRNA Details
| Gene Symbol: Zcchc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026479.4 | NP_080755.2 | 1, 2 | 62 | ||
| GenBank mRNA | AK010803.1 | - | 1, 2 | 73 | ||
| AK020177.1 | - | 1, 2 | 63 | |||
| AK027985.1 | - | 1, 2 | 70 | |||
| AK169856.1 | - | 1, 2 | 63 | |||
| BC025078.1 | - | 1, 2 | 78 | 4504126 | ||
| BC087902.1 | - | 1, 2 | 62 | 30505854 | ||

Zcchc10
Genomic Map