siRNA Details
| Gene Symbol: Ooep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026480.3 | NP_080756.1 | 2 | 321 | ||
| GenBank mRNA | AB283026.1 | - | 2 | 261 | ||
| AK010266.1 | - | 2 | 316 | |||
| AK010809.1 | - | 2 | 322 | |||
| AK133554.1 | - | 2 | 286 | |||
| AK135950.1 | - | 2 | 287 | |||
| AK136199.1 | - | 287 | ||||
| AK145510.1 | - | 2 | 287 | |||
| BC062245.1 | - | 2 | 272 | 3470921 | ||
| DQ238107.1 | - | 2 | 322 | |||

Ooep
Genomic Map