siRNA Details
| Gene Symbol: Tctn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026486.3 | NP_080762.1 | 7 | 918 | ||
| GenBank mRNA | AK012720.1 | - | 7 | 892 | ||
| AK014476.1 | - | 7 | 884 | |||
| AK031829.1 | - | 7 | 919 | |||
| AK032378.1 | - | 6 | 1003 | |||
| AK087856.1 | - | 7 | 877 | |||
| AK162182.1 | - | 676 | ||||
| AK205474.1 | - | 2 | 156 | |||
| BC058375.1 | - | 7 | 781 | 6828178 | ||
| DQ278869.1 | - | 7 | 916 | |||

Tctn2
Genomic Map