siRNA Details
| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026499.4 | NP_080775.3 | 4 | 704 | ||
| GenBank mRNA | AK004831.1 | - | 4 | 685 | ||
| AK012605.1 | - | 4 | 452 | |||
| AK051608.1 | - | 2 | 857 | |||
| AK133872.1 | - | 4 | 685 | |||
| AK146293.1 | - | 4 | 682 | |||
| AK159519.1 | - | 4 | 682 | |||
| AK171709.1 | - | 3 | 1968 | |||
| AK179065.1 | - | 3 | 333 | |||
| AK185344.1 | - | 1 | 152 | |||
| AK208133.1 | - | 1 | 151 | |||
| BC012039.1 | - | 4 | 691 | 2648111 | ||

Srsf6
Genomic Map