siRNA Details
| Gene Symbol: Pdzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081064.1 | NP_001074533.1 | 2 | 669 | ||
| GenBank mRNA | AK020883.1 | - | 3 | 266 | ||
| AK159017.1 | - | 4 | 1427 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzd2| Gene Symbol: Pdzd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081064.1 | NP_001074533.1 | 2 | 669 | ||
| GenBank mRNA | AK020883.1 | - | 3 | 266 | ||
| AK159017.1 | - | 4 | 1427 | |||