siRNA Details
| Gene Symbol: Smarcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114096.1 | NP_001107568.1 | 12, 13 | 1225 | ||
| NM_001114097.1 | NP_001107569.1 | 12, 13 | 1225 | |||
| NM_198160.2 | NP_937803.1 | 12, 13 | 1225 | |||
| GenBank mRNA | AK089461.1 | - | 12, 13 | 1161 | ||
| AK146971.1 | - | 12, 13 | 1154 | |||
| AK170530.1 | - | 12, 13 | 1160 | |||
| BC040363.1 | - | 12, 13 | 1140 | |||
| BC053452.1 | - | 3, 4 | 286 | |||
| BC058720.1 | - | 12, 13 | 1167 | 5705438 | ||
| BC062102.1 | - | 1, 2 | 27 | |||

Smarcc2
Genomic Map